Learn More
Cayman Chemical AcesulfamepotasIum salt 100g
SDP
Supplier:  Cayman Chemical 33356100 G
Encompass_Preferred
A non-caloric sweetener; has been found in wastewater and is considered a pollutant
Catalog No. 50-294-6694