Learn More
Medchemexpress LLC Acetamide, 2-[2-(2-azidoethoxy)ethoxy]-N-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl] | 2267306-14-7 | 96.6% | 444.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137537100MG
Encompass_Preferred
Pomalidomide-PEG2-azide is a synthesized E3 ligase ligand-linker conjugate, incorporating a Pomalidomide-based cereblon ligand and a 2-unit PEG linker, for use in PROTAC technology. It functions as a click chemistry reagent, with an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide-based cereblon ligand and 2-unit PEG linker
  • Used in PROTAC technology
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions
  • Participates in SPAAC reactions
  • For long-term storage, store at -20°C, protected from light
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-51811