Learn More
Medchemexpress LLC Acetamide 2-(2-aminoethoxy)-N-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]HCl | 2380273-67-4 | 97.1% | 410.81 g/mol | C17H19ClN4O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1367725MG
Encompass_Preferred
Pomalidomide-PEG1-NH2 hydrochloride is a pomalidomide-based E3 ligase ligand-linker conjugate (hydrochloride salt) featuring a short PEG linker terminated in a primary amine. It is designed for use in targeted protein degradation chemistry as a cereblon-recruiting ligand and for covalent coupling to target ligands via the terminal amine.
  • Primary amine-terminated PEG1 linker for conjugation to carboxyl groups.
  • E3 ligase (cereblon) recruiter based on a pomalidomide scaffold.
  • Supplied as a hydrochloride salt for improved stability and handling.
  • High purity (~97%) suitable for research and medicinal chemistry.
  • Available in multiple small-mass package sizes for synthesis work.
Catalog No. 50-002-21842