Learn More
Medchemexpress LLC Acetamide, 2,2-dichloro-N-(2,6-dimethylphenyl)- | 39084-88-3 | 99.9% | 232.11 | 5 MG

Supplier: Medchemexpress LLC HY440095MG
Lidocaine impurity 1 is an impurity of Lidocaine, intended for research use only. This product is not sold to patients.
- Molecular weight: 232.11
- Formula: C10H11Cl2NO
- CAS No.: 39084-88-3
- Appearance: Solid
- SMILES: O=C(NC1=C(C)C=CC=C1C)C(Cl)Cl
- Purity: 99.92%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.