Learn More
Medchemexpress LLC Acetamide, 2-[(8-ethyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)thio]-N-(2-methylphenyl)- | 3070282-38-8 | 99.05% | 377.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17647150MG
Encompass_Preferred
IGF2BP3-IN-1 (Compound I3IN-122) is an inhibitor of IGF2BP3, an RNA binding protein, with an IC50 of > 50 μM. It can be used for cancer research. This product is for research use only and is not sold to patients.
  • Inhibitor of IGF2BP3 (a RNA binding protein)
  • Can be used for cancer research
  • Purity: 99.05%
  • Available in various quantities
Catalog No. 50-003-86973