Learn More
Medchemexpress LLC Acetaminophen-13C2,15N | 360769-21-7 | 98.3% | C613C2H915NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY66005S45MG
Encompass_Preferred
Acetaminophen-13C2,15N is the 13C and 15N labeled version of Acetaminophen. It is a selective cyclooxygenase-2 (COX-2) inhibitor, widely used as an antipyretic and analgesic agent. It also acts as a potent inhibitor of hepatic N-acetyltransferase 2 (NAT2).
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitative analysis during drug development
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34779