Learn More
Medchemexpress LLC Acetaminophen-d3 | 60902-28-5 | >99.9% | C8H6D3NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY66005S11MG
Encompass_Preferred
Acetaminophen-d3 is the deuterium labeled form of Acetaminophen, a selective cyclooxygenase-2 (COX-2) inhibitor and a widely used antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34351