Learn More
Cayman Chemical AcetamInophen-d4 50mg
SDP
Supplier:  Cayman Chemical 1824550 MG
Encompass_Preferred
An internal standard for the quantification of acetaminophen by GC- or LC-MS
Catalog No. 50-297-9678