Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetaminophen-d4 | 64315-36-2 | 99.6% | 155.19 | C8H5D4NO2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY66005S10MG

Encompass_Preferred

Acetaminophen-d4 is a deuterium-labeled isotopologue of acetaminophen supplied for analytical and research applications, particularly as an internal standard in mass spectrometry and pharmacokinetic studies. It is provided as a high-purity solid suitable for quantitative assays.

  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High purity (99.6%) for analytical reliability.
  • Molecular formula C8H5D4NO2 and molecular weight 155.19 for accurate mass calibration.
  • Supplied as a solid powder in small vial sizes (e.g., 10 MG) for trace analysis.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).

Catalog No. 50-002-97913


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.