Learn More
Medchemexpress LLC Acetaminophen-d4 | 64315-36-2 | 99.6% | C8H5D4NO2 | 50 MG
Supplier: Medchemexpress LLC HY66005S50MG
Acetaminophen-d4 is the deuterium labeled form of Acetaminophen (Paracetamol). Acetaminophen is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM, widely recognized as an antipyretic and analgesic agent. It is also noted as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
- Serves as a deuterium-labeled variant of acetaminophen
- Can be utilized as a tracer in various applications
- Suitable for use as an internal standard for quantitative analysis methods
- Incorporation of stable heavy isotopes is valuable for quantitation during drug development