Learn More
Medchemexpress LLC Acetaminophen-d7 | 1219798-53-4 | 99.92% | C8H2D7NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY66005S25MG
Encompass_Preferred
Acetaminophen-d7 is the deuterium-labeled Acetaminophen. Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM and is a widely used antipyretic and analgesic agent. It is also a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33712