Learn More
Medchemexpress LLC Acetaminophen-d7 | 1219798-53-4 | MFCD08705146 | 99.9% | 158.21 g/mol | C8H2D7NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY66005S210MG
Encompass_Preferred
Acetaminophen-d7 is a deuterium-labeled analogue of acetaminophen used as an internal standard and tracer in mass spectrometry and metabolic studies. The compound contains seven deuterium atoms (d7) for unambiguous mass differentiation and is supplied as a high-purity reference suitable for quantitative LC-MS/MS analyses.
  • Deuterium-labeled internal standard for mass spectrometry
  • High isotopic enrichment for clear mass separation
  • Suitable for LC-MS/MS and quantitative analyses
  • Compact 10 mg quantity for analytical workflows
  • High purity for reproducible analytical results
Catalog No. 50-002-99116