Learn More
Cayman Chemical AcetamIprId 10mg
SDP
Supplier:  Cayman Chemical 2412910 MG
Encompass_Preferred
A neonicotinoid insecticide; an agonist of insect nAChRs; activates nAChRs containing N. lugens α1 and rat β2 subunits (EC50 = 67 µM in X. laevis oocytes); leads to sexual and aggressive behaviors in adult male offspring after administration to pregnant mice (1 mg/kg); decreases anxiety-like behavior of adult male offspring in the light-dark transition test (1 and 10 mg/kg)
Catalog No. 50-293-3129