Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetanilide NISTR SRMR141E
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST141E
Encompass
This Standard Reference Material (SRM) is intended for use in validating microchemical procedures for the This Standard Reference Material (SRM) is intended for use in validating microchemical procedures for the determination of carbon hydrogen and nitrogen in organic matter. A unit of SRM 141e contains 2 g of crystalline material.
Catalog No. 50-290-7955