Learn More
Teknova Acetate and Sucrose Buffer, pH 5.5, Endotoxin Tested, 45mL, 25 Tubes per Pack, Sterile. 2 pack.
SDP
Supplier:  Teknova S85022PK
Encompass
2.72g/L Sodium Acetate, 90 g/L Sucrose
Catalog No. 50-238-7285