Learn More
Apexbio Technology LLC Acetate gossypol 12542-36-8 50mg
SDP
Supplier:  Apexbio Technology LLC N213450
Encompass_Preferred
Acetate gossypol (CAS 12542-36-8) is a polyphenolic compound isolated from the seeds and bark of Gossypium herbaceum L It functions primarily as an inhibitor of anti-apoptotic Bcl-2 family proteins thereby promoting apoptosis in various cancer cell types By disrupting mitochondrial membrane potential and modulating cell survival pathways acetate gossypol has been utilized in studies investigating mechanisms of programmed cell death and antitumor activity This compound is frequently employed as a molecular tool in cancer biology and apoptosis research Store under sealed cool and dry conditions to ensure stability
Catalog No. 50-224-0647