Learn More
Medchemexpress LLC Acetic acid, 2-[2-[2-[[(4-methylphenyl)sulfonyl)oxy]ethoxy]ethoxy]-, 1,1-dimethylethyl ester | 882518-89-0 | 97.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13053250MG
Encompass_Preferred
Tos-PEG2-CH2-Boc is a PROTAC linker that belongs to the PEG class. It is utilized in the synthesis of PROTAC molecules. PROTACs themselves consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. These molecules harness the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • Classified as a PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTAC molecules.
  • For scientific research or drug registration purposes only and not for personal use.
Catalog No. 50-003-38750