Learn More
Medchemexpress LLC Acetic acid, 2-bromo-, 2,5-dioxo-1-pyrrolidinyl ester | 42014-51-7 | MFCD00058571 | 99.2% | 236.02 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW012166
Encompass_Preferred
N-Succinimidyl bromoacetate is a heterobifunctional cross-linking reagent. It can be used to prepare peptide-protein immunogens.
  • Heterobifunctional cross-linking reagent.
  • For research use only.
Catalog No. 50-004-51308