Learn More
Medchemexpress LLC Acetic acid, 2-(triphenylphosphoranylidene)-, methyl ester | 2605-67-6 | ≥98.0% | C21H19O2P | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY77175
Encompass_Preferred
Methyl 2-(triphenylphosphoranylidene)acetate is a biochemical reagent and pharmaceutical intermediate. It is primarily used in life science research. It can also serve as an intermediate to construct carbon-carbon double bond skeletons and synthesize other active compounds.
  • Used to construct carbon-carbon double bond skeletons
  • Synthesizes other active compounds
Catalog No. 50-003-58655