Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetic acid glacial ACS re
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6950924X4L
Encompass
Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in SOLUTIONon and gaseous commonly referred as phases. It is weakly acidic in nature.
Catalog No. 50-290-3343