Learn More
ATCC Bacteroides fragilis
Supplier:  ATCC 23745
Encompass
Bacteroides fragilis strain [ICPB 3498, NCTC l0581] is a bacterial strain isolated from pleural fluid. This product has applications in media testing and quality control.
Catalog No. 50-238-0188