Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone BioRenewable ACS r200L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 904082
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is 100% biorenewable and thus aligns with Safer Solvents and Auxiliaries and Use of Renewable Feedstocks. Click here for more information.This ACS grade Acetone is a drop-in replacement for any Acetone application. Made from 100% renewable carbon (verified through C14 ASTM D6866-16 testing) and produced in an environmentally friendly way that reduces the carbon footprint of production and reliance on fossil-fuels. Because it is not from petroleum it is free from many non-renewable contaminants such as benzene phthalates and other phenols leading to higher quality product.
Catalog No. 50-290-6867
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.