Learn More
High Purity Products Acetone, MFCD00008765, 4L
Supplier:  High Purity Products 300445
Encompass_Preferred
Molecular Formula C3H6O, Molecular Weight 58.079, CMOS Grade, case of 4
Catalog No. NC9909295