Learn More
Supply Solutions Burdick & Jackson - Acetonitrile, BioSyn, Package Size: 20L, Container Type:steel NOWPAK, Suggested Applications: Biosynthesis, Applications requiring low water content
Supplier:  Supply Solutions BJ BN01720
Encompass_Preferred
75-05-8. Acetonitrile, BioSyn. Suggested Application: Biosynthesis, Applications requiring low water content. Package Size: 20L. Container type is steel NOWPAK. Hazard Class: 3. Package Group: II. Molecular Weight: 41.05. Density (g/mL) at 20deg C: 0.7822. Boiling point (deg C): 81.6. Freezing point (deg C): -43.8. Flashpoint (deg C): 6. Manufacturer Name: Burdick and Jackson. Manufacturer Part Number: BN017-20.; Size: 20L
Catalog No. 50-877-406