Learn More
Medchemexpress LLC Acetyl-11-keto-β-boswellic acid (Standard) | 67416-61-9 | 512.72 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0892R100MG
Encompass_Preferred
AKBA (Standard) is an analytical standard and active triterpenoid compound from the extract of Boswellia serrate. It is a novel Nrf2 activator intended for research and analytical applications.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC applications
  • Suitable for GC applications
  • Suitable for MS applications
Catalog No. 50-004-16897