Learn More
Medchemexpress LLC Acetyl CoA Carboxylase 1 (ACC1) Antibody | 266 kDa | 100 UL

Supplier: Medchemexpress LLC HYP80482100UL
Acetyl CoA Carboxylase 1 (ACC1) Antibody (YA646) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Acetyl CoA Carboxylase 1 (ACC1). It reacts with human and mouse samples and is suitable for Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P) applications.
- Supplied in 1*TBS (pH 7.4), 0.05% BSA, and 40% Glycerol
- Concentration of 1 mg/mL
- Preservative: 0.05% Sodium Azide
- Appearance: Liquid
- Shipping: With blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.