Learn More
Medchemexpress LLC Acetyl CoA Carboxylase 1 (ACC1) Antibody (YA646) | 266 kDa | 10 UL

Supplier: Medchemexpress LLC HYP8048210UL
Acetyl CoA Carboxylase 1 (ACC1) Antibody (YA646) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Acetyl CoA Carboxylase 1 (ACC1). This antibody is suitable for research use only and ships with blue ice, ensuring stability. It is formulated in TBS with BSA and Glycerol, including sodium azide as a preservative, and is stable for 1 year when stored at -20°C.
- Targets Acetyl CoA Carboxylase 1 (ACC1)
- Rabbit-derived IgG monoclonal antibody
- Non-conjugated and unmodified
- Reactive with human and mouse samples
- Applications include Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P)
- Predicted molecular weight of 266 kDa
- Protein A affinity purified
- Ships with blue ice for stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.