Learn More
Medchemexpress LLC Acetyl-DL-carnitine | 14992-62-2 | MFCD00871161 | 98.0% | 203.24 g/mol | C9H17NO4 | 50MG

Supplier: Medchemexpress LLC HY12635850MG
Acetyl-DL-carnitine (CAS 14992-62-2) is an endogenous metabolite provided as a white to off-white solid for research use. The product is supplied with supporting documentation for purity, solubility, storage, and handling for in vitro applications.
- High purity: 98.0%.
- Molecular weight 203.24 g/mol.
- Chemical formula C9H17NO4.
- Good aqueous solubility (≈250 mg/mL), may require ultrasonic treatment.
- Storage: store at 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
- Available in multiple pack sizes, including 50 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.