Learn More
Epigentek Inc Histone H3K14ac (Acetyl H3K14) Polyclonal Antibody (25 ul)
SDP
Supplier:  Epigentek Inc A4023025
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human Acetyl-Histone H3-K14. Tested Applications: WB, IHC, IF, IP, ChIP, ChIPseq. Reactivity: Broad Range, Mouse, Rat, Human. Alternative Names: H3K14ac antibody; H3K14a antibody; H3t; H3.4; H3/g; H3FT.
Catalog No. 50-109-5488