Learn More
ABclonal Technology IL23R Rabbit mAb
SDP
Supplier:  ABclonal Technology A2256120UL
Encompass_Preferred
The protein encoded by this gene is a subunit of the receptor for IL23A IL23 This protein pairs with the receptor molecule IL12RB1 IL12Rbeta1 and both are required for IL23A signaling This protein associates constitutively with Janus kinase 2 (JAK2) and also binds to transcription activator STAT3 in a ligand-dependent manner
Catalog No. 50-000-07683