Learn More
REVMAB BIOSCIENCES USA INC Acetyl-Histone H3 (Lys4) [RM149], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31106300
Encompass_Preferred
Anti-Acetyl-Histone H3 (Lys4) rabbit monoclonal antibody, Clone RM149 (100UG); WB, ELISA, Multiplex, ICC validated; Product info: www.revmab.com
Catalog No. NC1729093