Learn More
Cayman Chemical Acetyl PentapeptIde-1 10mg
SDP
Supplier:  Cayman Chemical 2726110 MG
Encompass_Preferred
A pentapeptide; decreases IL-8 secretion in human keratinocytes when used in combination with acetyl hexapeptide-36 and acetyl hexapeptide-38
Catalog No. 50-294-1528