Learn More
Selleck Chemical LLC Acetyl phosphate E5841-25MG
SDP
Supplier:  Selleck Chemical LLC E584125MG
Encompass_Preferred
Acetyl phosphate(lithium potassium) is an endogenous metabolite bypasses the LytS membrane potential sensor by phosphorylating LytR twice as fast influencing lrgAB operon regulation in glucose-rich conditions without membrane potential disruption Lithium potassium acetyl phosphate is studied for its electrochemical properties and role in phosphorus chemistry with potential applications in lithium-ion battery technology and catalytic processes
Catalog No. 50-000-41694