Learn More
Cayman Chemical Acetyl Podocarpic Acid Anhydr
SDP
Supplier:  Cayman Chemical 100076865 MG
Encompass_Preferred
A potent, semisynthetic LXR agonist derived from extracts of the mayapple; induces the expression of the ABCA1 reverse cholesterol transporter to increase the efflux of cholesterol from enterocytes and thus inhibits the overall absorption of cholesterol (ED50 = 1 nM)
Catalog No. 50-289-1340