Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 98.97% | 402.48 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01098110MM/1ML
Encompass_Preferred
Acetyl tributyl citrate (Tributyl O-acetylcitrate) is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It can be used in cosmetics, food packaging, and as a flavoring substance for food. For research use only.
  • Used as a pharmaceutical excipient
  • Biodegradable hydrophobic plasticizer
  • Suitable for cosmetics applications
  • Can be used in food packaging
  • Acts as a flavoring substance for food
  • For research use only
Catalog No. 50-004-51704