Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 99.0% | 402.48 | 1 MG
Supplier: Medchemexpress LLC HYW010981S1MG
Acetyl tributyl citrate is a pharmaceutical excipient and a biodegradable hydrophobic plasticizer. It is suitable for use in cosmetics, food packaging, and as a food flavoring substance. This product is for research use only and not intended for patient use.
- Functions as a pharmaceutical excipient
- Biodegradable hydrophobic plasticizer
- Applicable in cosmetics and food packaging
- Can be used as a food flavoring substance
- Rapidly metabolized in rat and human plasma and liver microsomes
- Metabolized by multiple human CYP isoforms, including CYP2C19 and CYP3A4
- Demonstrates rapid absorption and elimination in vivo, with 27.4% oral bioavailability in rats