Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 99.0% | 402.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0109815MG
Encompass_Preferred
Acetyl tributyl citrate, also known as Tributyl O-acetylcitrate, is a pharmaceutical excipient and a biodegradable hydrophobic plasticizer. It is rapidly metabolized in rat plasma, rat and human liver microsomes, with significant activity from CYP2C19 and CYP3A4. It also shows rapid absorption and elimination in rats with an oral bioavailability of 27.4%.
  • Utilized in cosmetics
  • Used in food packaging
  • Functions as a flavoring substance for food
Catalog No. 50-004-52948