Learn More
Medchemexpress LLC Acetyl tributyl citrate (Standard) | 77-90-7 | 98.77% | 402.48 | 50 MG
Supplier: Medchemexpress LLC HYW010981R50MG
Acetyl tributyl citrate (Standard) is the analytical standard for Acetyl tributyl citrate, also known as Tributyl O-acetylcitrate or ATBC. It functions as an alternative plasticizer and is intended for various research and analytical applications.
- Used for research purposes.
- Serves as an analytical standard.
- Functions as an alternative plasticizer.
- Employed in qualitative, quantitative, and methodological research experiments.
- Suitable for techniques like HPLC, GC, and MS.