Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylarenobufagin | 184673-79-8 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN69055MG

Encompass_Preferred

Acetylarenobufagin is a steroidal hypoxia inducible factor-1 (HIF-1) modulator, intended for research use only.

  • Targets HIF-1
  • Purity of 98.63%
  • Molecular weight of 458.54
  • Formula is C26H34O7
  • Appears as a white to off-white solid
  • Initial source is animals, toad

Catalog No. 50-004-19342


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.