Learn More
Apexbio Technology LLC (±)-Acetylcarnitine chloride 2504-11-2 100mg
SDP
Supplier:  Apexbio Technology LLC B6273100
Encompass_Preferred
( )-Acetylcarnitine chloride (CAS 2504-11-2) is a cholinergic agonist and a metabolic intermediate involved in lipid metabolism It interacts with acetylcholine receptors including nicotinic and muscarinic subtypes and demonstrates the ability to modulate neurotransmitter systems In retinal ganglion cells both acetylcarnitine and acetylcholine suppress GABAergic responses indicating an influence on inhibitory synaptic transmission In animal models acetylcarnitine derivatives reduce ventricular arrhythmia following coronary artery ligation and enhance mitochondrial function Additionally acetyl-L-carnitine exhibits antinociceptive effects dependent on activation of the phospholipase C-IP3 signaling pathway This compound is frequently utilized in studies of neuromodulation cardiac function and pain signaling mechanisms
Catalog No. 50-000-87903