Learn More
Medchemexpress LLC Acetylcholine (chloride) | 60-31-1 | 99.9% | 10 G

Supplier: Medchemexpress LLC HYB028210G
Acetylcholine chloride (ACh chloride) is a neurotransmitter, a potent and BBB-permeable cholinergic agonist. It modulates the activity of dopaminergic (DAergic) neurons by stimulating nicotinic acetylcholine receptors (nAChRs) and inhibits p53 mutant peptide aggregation in vitro.
- Modulates activity of dopaminergic neurons
- Inhibits p53 mutant peptide aggregation
- Targets nAChR, calcium channel, and endogenous metabolite
- Suitable for neurological disease and cancer research
- Molecular Weight: 181.66
- Formula: C7H16ClNO2
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.