Learn More
Medchemexpress LLC Acetylcholine chloride | 60-31-1 | 99.91% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB02825G
Encompass_Preferred
Acetylcholine chloride (ACh chloride) is a potent, BBB-permeable cholinergic agonist and neurotransmitter. It modulates the activity of dopaminergic neurons by stimulating nicotinic acetylcholine receptors (nAChRs) and inhibits p53 mutant peptide aggregation in vitro.
  • Modulates activity of dopaminergic neurons
  • Stimulates nicotinic acetylcholine receptors (nAChRs)
  • Inhibits p53 mutant peptide aggregation in vitro
  • BBB-permeable cholinergic agonist
Catalog No. 50-004-06034