Learn More
Medchemexpress LLC Acetylcholine chloride | 60-31-1 | MFCD00011698 | 99.91% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB028210MM/1ML
Encompass_Preferred
Acetylcholine chloride (ACh chloride) is a neurotransmitter and a potent, BBB-permeable cholinergic agonist. It modulates the activity of dopaminergic (DAergic) neurons by stimulating nicotinic acetylcholine receptors (nAChRs) and has been shown to inhibit p53 mutant peptide aggregation in vitro.
  • Neurotransmitter and cholinergic agonist
  • BBB-permeable
  • Modulates dopaminergic (DAergic) neurons
  • Stimulates nicotinic acetylcholine receptors (nAChRs)
  • Inhibits p53 mutant peptide aggregation in vitro
Catalog No. 50-004-09020