Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylcholine-d4 chloride | 344298-94-8 | MFCD00039479 | 98.0% | 185.69 g/mol | C7H12D4ClNO2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0282S10MG

Encompass_Preferred

Acetylcholine-d4 chloride is the deuterium-labeled (d4) stable isotope of acetylcholine chloride intended for research and analytical applications. It is typically used as an internal standard or tracer in mass spectrometry and isotope-labeling studies, and is supplied as a solid with high isotopic enrichment and reported purity of 98.0%.

  • Deuterium-labeled stable isotope suitable for analytical use.
  • Used as an internal standard and tracer in mass spectrometry.
  • Reported purity 98.0%.
  • Molecular weight 185.69 g/mol.
  • CAS number 344298-94-8.

Catalog No. 50-002-97219


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.