Learn More
Medchemexpress LLC Acetylcholine-d9 (chloride) | 344298-95-9 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0282S15MG
Encompass_Preferred
Acetylcholine-d9 (chloride) is a deuterium-labeled Acetylcholine chloride, a neurotransmitter and a potent, BBB-permeable cholinergic agonist. It modulates dopaminergic neuron activity by stimulating nicotinic acetylcholine receptors (nAChRs) and inhibits p53 mutant peptide aggregation in vitro.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06015