Learn More
Selleck Chemical LLC Acetylcysteine (N-acetylcysteine) S1623-5g
SDP
Supplier:  Selleck Chemical LLC S16235G
Encompass_Preferred
Acetylcysteine (N-acetyl-l-cysteine NAC N-acetylcysteine) is a ROS(reactive oxygen species) inhibitor that antagonizes the activity of proteasome inhibitors It is also a tumor necrosis factor production inhibitor Acetylcysteine(N-acetyl-l-cysteine) suppresses TNF-induced NF- B activation through inhibition of I B kinases Acetylcysteine(N-acetyl-l-cysteine) induces apoptosis via the mitochondria-dependent pathway Acetylcysteine(N-acetyl-l-cysteine) inhibits ferroptosis and virus replication Solutions are unstable and should be fresh-prepared
Catalog No. 50-310-8683