Learn More
Medchemexpress LLC Acetylcysteine | 616-91-1 | 99.9% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB021550G
Encompass_Preferred
Acetylcysteine (N-Acetylcysteine) is a mucolytic agent that has various biological activities. It can effectively reduce the thickness of mucus and is known to cross the blood-brain barrier.
  • Reduces the thickness of mucus
  • Acts as an inhibitor of reactive oxygen species (ROS)
  • Functions as a precursor to cysteine
  • Prevents hemin-induced ferroptosis by neutralizing toxic lipids generated by the arachidonate-dependent activity of 5-lipoxygenases
  • Induces cell apoptosis
  • Demonstrates anti-influenza virus activities
  • Represents the most stable form of cysteine for drug delivery
  • Can be utilized in disulfidptosis studies
Catalog No. 50-004-06401