Learn More
Medchemexpress LLC Acetylcysteine-d3 | 131685-11-5 | 97.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0215S5MG
Encompass_Preferred
Acetylcysteine-d3 is the deuterium-labeled version of Acetylcysteine. It functions as a mucolytic agent by reducing mucus thickness and is a ROS inhibitor. It also acts as a cysteine precursor, preventing hemin-induced ferroptosis through neutralizing toxic lipids. Acetylcysteine induces cell apoptosis and exhibits anti-influenza virus properties. It is considered the most stable form of cysteine for drug delivery and can be utilized in disulfidptosis studies.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05786