Learn More
Abcam Acetylcysteine (N-Acetyl-L-cysteine), ROS inhibitor, 50MG
Supplier:  Abcam AB14303250MG
Encompass_Preferred
MW 163.2 Da, Purity >99%. ROS inhibitor. L-cysteine prodrug. Mucolytic agent. Inhibits viral stimulation by reactive oxygen intermediates. Shows mucolytic, hepatoprotective and antiviral effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3302