Learn More
Medchemexpress LLC Acetylene-linker-Val-Cit-PABC-MMAE | 1411977-95-1 | 95.2% | 1307.62 | C67H106N10O16 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY198125MG
Encompass_Preferred
Acetylene-linker-Val-Cit-PABC-MMAE is an alkyne-functionalized antibody-drug conjugate (ADC) drug-linker conjugate combining a Val-Cit-PABC cleavable linker with the tubulin inhibitor MMAE. It is designed for ADC synthesis and for use in copper-catalyzed azide-alkyne cycloaddition (CuAAC) click-chemistry applications.
  • Alkyne functional group for CuAAC click chemistry.
  • Val-Cit-PABC cleavable linker enables protease-mediated payload release.
  • Contains MMAE tubulin inhibitor payload for ADC research.
  • High purity suitable for research applications.
  • Available in small research quantities for conjugation workflows.
Catalog No. 50-002-03293